3-(1-Naphthyloxy)benzonitrile
Catalog # APOSOR470291-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:245.275 g/mol
Storage Temperature:Ambient
CAS Number:1314406-41-1
Formula:C₁₇H₁₁NO
MDL Number:MFCD19442773