Order Entry
Export
ContactUsLinkComponent
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid
Catalog # APOSOR470236-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid
Catalog # APOSOR470236-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :246.262 g/mol
Storage Temperature:Ambient
CAS Number:1272755-83-5
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD18651747