Order Entry
Export
ContactUsLinkComponent
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
Catalog # APOSOR470228-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
Catalog # APOSOR470228-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :241.288 g/mol
Storage Temperature:Ambient
CAS Number:1272755-94-8
Formula:C₁₄H₁₅N₃O
MDL Number:MFCD18651735