Order Entry
Export
ContactUsLinkComponent
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid
Catalog # APOSOR470226-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid
Catalog # APOSOR470226-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :246.262 g/mol
Storage Temperature:Ambient
CAS Number:1272756-14-5
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD18651731