Order Entry
Export
ContactUsLinkComponent
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate
Catalog # APOSOR470225-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate
Catalog # APOSOR470225-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :274.315 g/mol
Storage Temperature:Ambient
CAS Number:1272756-22-5
Formula:C₁₅H₁₈N₂O₃
MDL Number:MFCD18651730