2,6-Bis[1-[(2,6-dimethylphenyl)imino]ethyl]pyridine
Catalog # APOSOR470167-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:369.502 g/mol
Storage Temperature:Ambient
CAS Number:204203-16-7
Formula:C₂₅H₂₇N₃
MDL Number:MFCD18375270