Order Entry
Export
ContactUsLinkComponent
 
Ethyl 5,8-Dichloroimidazo[1,5-a]pyrazine-1-carboxylate
Catalog # APOSOR470166-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 5,8-Dichloroimidazo[1,5-a]pyrazine-1-carboxylate
Catalog # APOSOR470166-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.077 g/mol
Storage Temperature:Ambient
CAS Number:1256633-28-9
Formula:C₉H₇Cl₂N₃O₂
MDL Number:MFCD18071050