N-(2,5-Dimethyl-4-nitrophenyl)acetamide
Catalog # APOSOR470041-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:208.214 g/mol
Storage Temperature:Ambient
CAS Number:6954-69-4
Formula:C₁₀H₁₂N₂O₃
MDL Number:MFCD00547733