(6S)-2-Nitro-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazin-6-ol 97%
Catalog # APOSOR46771-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:185.137 g/mol
Storage Temperature:Ambient
CAS Number:187235-08-1
Formula:C₆H₇N₃O₄
MDL Number:MFCD08436173