
7-Bromo-4,6-dichloroquinazoline
Catalog # APOSOR46571-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.933 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1260847-61-7
Formula:C₈H₃BrCl₂N₂
MDL Number:MFCD13193278