Order Entry
Export
ContactUsLinkComponent
 
Irritant
6-Bromo-1H-imidazo[4,5-c]pyridine
Catalog # APOSOR46517-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-1H-imidazo[4,5-c]pyridine
Catalog # APOSOR46517-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.02 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1312440-90-6
Formula:C₆H₄BrN₃
MDL Number:MFCD19288874