Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine
5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1245645-07-1
:
Formula:
C₆H₅IN₄
MDL Number:
MFCD18072706
:
260.035 g/mol
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III