Order Entry
Export
ContactUsLinkComponent
2,3-Butylene carbonate
Irritant
2,3-Butylene carbonate
Catalog # APOSOR46258-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Butylene carbonate
Catalog # APOSOR46258-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :116.115 g/mol
Boiling Pt:85 °C (0.06 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:4437-70-1
Formula:C₅H₈O₃
MDL Number:MFCD00060135