Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-b-Amino-3,4-dichlorobenzenepropanol
(S)-b-Amino-3,4-dichlorobenzenepropanol 97%
(S)-b-Amino-3,4-dichlorobenzenepropanol 97%
(S)-b-Amino-3,4-dichlorobenzenepropanol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
(S)-b-Amino-3,4-dichlorobenzenepropanol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
325687-06-7
:
Formula:
C₉H₁₁Cl₂NO
MDL Number:
MFCD17214639
:
220.096 g/mol
Storage Temperature:
Ambient