Order Entry
Export
ContactUsLinkComponent
5,5'-Disulphanediylbis(1-phenyl-1H-tetrazole) 95%
Irritant
5,5'-Disulphanediylbis(1-phenyl-1H-tetrazole) 95%
Catalog # APOSOR45146-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5,5'-Disulphanediylbis(1-phenyl-1H-tetrazole) 95%
Catalog # APOSOR45146-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :354.413 g/mol
Melting Pt:145 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:517-07-7
Formula:C₁₄H₁₀N₈S₂
MDL Number:MFCD00134361