Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethylpiperidine
2,4-Dimethylpiperidine
2,4-Dimethylpiperidine
:
Apollo Scientific
2,4-Dimethylpiperidine
:
Apollo Scientific
:
6287-19-0
:
:
C₇H₁₅N
:
MFCD00129009
:
113.201 g/mol
:
GHS07
:
Ambient