
4-Boc-6-oxo-1,4-oxazepane
Catalog # APOSOR451216-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.246 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:748805-97-2
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD11976154