Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%
Irritant
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
66500-55-8
:
Formula:
C₉H₁₄O₄
MDL Number:
MFCD11847571
:
186.205 g/mol
:
GHS07
Storage Temperature:
Ambient