1,4-Dioxaspiro[4.5]decane-8-acetic acid
Catalog # APOSOR451153-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:200.232 g/mol
Storage Temperature:Ambient
CAS Number:134136-04-2
Formula:C₁₀H₁₆O₄
MDL Number:MFCD16620931