Order Entry
Export
ContactUsLinkComponent
8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane
8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane
Catalog # APOSOR451151-1G
Supplier:  Apollo Scientific
CAS Number:  
8-(Iodomethyl)-1,4-dioxaspiro[4.5]decane
Catalog # APOSOR451151-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :282.119 g/mol
Storage Temperature:Ambient
CAS Number:625114-56-9
Formula:C₉H₁₅IO₂
MDL Number:MFCD26387804