β-Oxo-cyclobutanepropanoic acid 1,1-dimethylethyl ester
Catalog # APOSOR451076-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.259 g/mol
Storage Temperature:Ambient
CAS Number:134302-09-3
Formula:C₁₁H₁₈O₃
MDL Number:MFCD11974506