4-Thioxo-1,3-thiazolidin-2-one
Catalog # APOSOR450087-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:133.192 g/mol
Storage Temperature:Ambient
CAS Number:4303-29-1
Formula:C₃H₃NOS₂
MDL Number:MFCD02167269