(5Z)-5-(Ethoxymethylene)-2-thioxo-1,3-thiazolidin-4-one
Catalog # APOSOR450084-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189.255 g/mol
Storage Temperature:Ambient
CAS Number:86240-28-0
Formula:C₆H₇NO₂S₂
MDL Number:MFCD00175004