Order Entry
Export
ContactUsLinkComponent
Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate 95%
Irritant
Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate 95%
Catalog # APOSOR43693-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate 95%
Catalog # APOSOR43693-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :224.276 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1414377-89-1
Formula:C₁₁H₁₂O₃S
MDL Number:MFCD28347696