Order Entry
Export
ContactUsLinkComponent
5-Bromo-2,4-dichlorotoluene 98%
5-Bromo-2,4-dichlorotoluene 98%
Catalog # APOSOR43684-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2,4-dichlorotoluene 98%
Catalog # APOSOR43684-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.925 g/mol
Storage Temperature:Ambient
CAS Number:85072-41-9
Formula:C₇H₅BrCl₂
MDL Number:MFCD09800823