Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2,5-Dichlorophenyl)methanesulphonyl chloride
(2,5-Dichlorophenyl)methanesulphonyl chloride
(2,5-Dichlorophenyl)methanesulphonyl chloride
:
Apollo Scientific
(2,5-Dichlorophenyl)methanesulphonyl chloride
:
Apollo Scientific
:
163295-71-4
:
:
C₇H₅Cl₃O₂S
:
MFCD11976103
:
8,III
:
259.537 g/mol
:
3261
:
GHS05
:
Ambient