Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chlorothiazolo[5,4-b]pyridin-2-amine
5-Chlorothiazolo[5,4-b]pyridin-2-amine 98%
5-Chlorothiazolo[5,4-b]pyridin-2-amine 98%
:
Apollo Scientific
5-Chlorothiazolo[5,4-b]pyridin-2-amine 98%
:
Apollo Scientific
:
31784-71-1
:
:
C₆H₄ClN₃S
:
MFCD00275399
:
185.634 g/mol
:
GHS07
:
Ambient