7-Bromo-4-chloro-2,8-dimethylquinoline
Catalog # APOSOR42039-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.553 g/mol
Storage Temperature:Ambient
CAS Number:1189106-62-4
Formula:C₁₁H₉BrClN
MDL Number:MFCD12675042