Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
:
Apollo Scientific
6-Bromo-3-methyl-[1,2,4]triazolo[4,3-a]pyridine
:
Apollo Scientific
:
108281-78-3
:
:
C₇H₆BrN₃
:
MFCD08276188
:
212.047 g/mol
:
147…149 °C
:
GHS07
:
Ambient