0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
3-Bromopiperidine-2,6-dione
3-Bromopiperidine-2,6-dione 95%
3-Bromopiperidine-2,6-dione 95%
:
Apollo Scientific
3-Bromopiperidine-2,6-dione 95%
:
Apollo Scientific
:
62595-74-8
:
:
C₅H₆BrNO₂
:
MFCD11053059
:
192.011 g/mol
:
107 °C
:
GHS07
:
Ambient