
N-(3,4-Dimethylbenzyl)cyclopropylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1257535-33-3
:
Formula:C₁₂H₁₇N·ClH
MDL Number:MFCD17168269
ADR:8,III
:211.731 g/mol
UN:1759
:GHS05
Storage Temperature:Refrigerator