Order Entry
Export
ContactUsLinkComponent
1,3-Dimethyl-1H-indazole-5-boronic acid
Irritant
1,3-Dimethyl-1H-indazole-5-boronic acid
Catalog # APOSOR40161-25MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dimethyl-1H-indazole-5-boronic acid
Catalog # APOSOR40161-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :190.007 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1310404-48-8
Formula:C₉H₁₁BN₂O₂
MDL Number:MFCD12028654