Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dimethyl-1H-indazole-4-boronic acid
1,5-Dimethyl-1H-indazole-4-boronic acid 95%
1,5-Dimethyl-1H-indazole-4-boronic acid 95%
Irritant
1,5-Dimethyl-1H-indazole-4-boronic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,5-Dimethyl-1H-indazole-4-boronic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1262512-81-1
:
Formula:
C₉H₁₁BN₂O₂
MDL Number:
MFCD12028651
:
190.007 g/mol
:
GHS07
Storage Temperature:
Refrigerator