0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Methoxy-3-methylbenzyl alcohol
2-Methoxy-3-methylbenzyl alcohol
2-Methoxy-3-methylbenzyl alcohol
:
Apollo Scientific
2-Methoxy-3-methylbenzyl alcohol
:
Apollo Scientific
:
74090-48-5
:
:
C₉H₁₂O₂
:
MFCD12031989
:
152.19 g/mol
:
GHS07
:
Ambient