Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-chlorobenzene-1,2-diamine
3-Bromo-4-chlorobenzene-1,2-diamine
3-Bromo-4-chlorobenzene-1,2-diamine
:
Apollo Scientific
3-Bromo-4-chlorobenzene-1,2-diamine
:
Apollo Scientific
:
1823624-85-6
:
:
C₆H₆BrClN₂
:
MFCD28154807
:
221.482 g/mol
:
GHS07
:
Ambient