Order Entry
Export
ContactUsLinkComponent
7-Cyclopropyl-2,7-diazaspiro[3.5]nonane dihydrochloride 95%
7-Cyclopropyl-2,7-diazaspiro[3.5]nonane dihydrochloride 95%
Catalog # APOSOR370123-250MG
Supplier:  Apollo Scientific
CAS Number:  
7-Cyclopropyl-2,7-diazaspiro[3.5]nonane dihydrochloride 95%
Catalog # APOSOR370123-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :239.185 g/mol
Storage Temperature:Ambient
CAS Number:1415562-71-8
Formula:C₁₀H₁₈N₂·2ClH
MDL Number:MFCD22124616