Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,3-Dihydroxybenzaldehyde ≥98%
4-Bromo-2,3-Dihydroxybenzaldehyde ≥98%
Catalog # APOSOR370094-500MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,3-Dihydroxybenzaldehyde ≥98%
Catalog # APOSOR370094-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :217.017 g/mol
Storage Temperature:Ambient
CAS Number:73275-98-6
Formula:C₇H₅BrO₃
MDL Number:MFCD30738024