Order Entry
Export
ContactUsLinkComponent
2-(2-Methoxyphenyl)-4,5-dihydrooxazole ≥98%
2-(2-Methoxyphenyl)-4,5-dihydrooxazole ≥98%
Catalog # APOSOR370071-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Methoxyphenyl)-4,5-dihydrooxazole ≥98%
Catalog # APOSOR370071-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.2 g/mol
Storage Temperature:Ambient
CAS Number:74272-88-1
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD22665893