Order Entry
Export
ContactUsLinkComponent
3-Ethyl-2,3-dimethyl-1,2-oxaziridine ≥98%
3-Ethyl-2,3-dimethyl-1,2-oxaziridine ≥98%
Catalog # APOSOR370058-25MG
Supplier:  Apollo Scientific
CAS Number:  
3-Ethyl-2,3-dimethyl-1,2-oxaziridine ≥98%
Catalog # APOSOR370058-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :101.147 g/mol
Storage Temperature:Ambient
CAS Number:3400-16-6
Formula:C₅H₁₁NO
MDL Number:MFCD18642810