Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Isopropyl-2-methyl-1,2-oxaziridine
3-Isopropyl-2-methyl-1,2-oxaziridine ≥98%
3-Isopropyl-2-methyl-1,2-oxaziridine ≥98%
APOSOR370057-500MG
:
Apollo Scientific
:
3-Isopropyl-2-methyl-1,2-oxaziridine ≥98%
APOSOR370057-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
101.147 g/mol
:
Ambient
:
1314904-03-4
:
C₅H₁₁NO
:
MFCD18642809