Order Entry
Export
ContactUsLinkComponent
N-(3,6-Dimethylpyridin-2-yl)pivalamide ≥98%
N-(3,6-Dimethylpyridin-2-yl)pivalamide ≥98%
Catalog # APOSOR370050-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(3,6-Dimethylpyridin-2-yl)pivalamide ≥98%
Catalog # APOSOR370050-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.284 g/mol
Storage Temperature:Ambient
CAS Number:1313762-34-3
Formula:C₁₂H₁₈N₂O
MDL Number:MFCD19443197