Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boric acid, pinacol ester
Boric acid, pinacol ester 97%
Boric acid, pinacol ester 97%
Boric acid, pinacol ester 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Boric acid, pinacol ester 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
25240-59-9
:
Formula:
C₆H₁₃BO₃
MDL Number:
MFCD15143603
:
143.977 g/mol
:
GHS07
:
GHS08
Storage Temperature:
Ambient