Order Entry
Export
ContactUsLinkComponent
6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine 97%
6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine 97%
Catalog # APOSOR350451-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine 97%
Catalog # APOSOR350451-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.632 g/mol
Storage Temperature:Ambient
CAS Number:1014720-73-0
Formula:C₉H₇ClN₄
MDL Number:MFCD14706108