Ethyl 6-oxo-3,6-dihydropyrimidine-4-carboxylate
Catalog # APOSOR350165-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.15 g/mol
Storage Temperature:Ambient
CAS Number:223788-14-5
:MFCD27665401
Formula:C₇H₈N₂O₃