2-Chloro-4,4'-bipyridine
Catalog # APOSOR350095-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:190.629 g/mol
Storage Temperature:Ambient
CAS Number:53344-73-3
Formula:C₁₀H₇ClN₂
MDL Number:MFCD11111098