Order Entry
Export
ContactUsLinkComponent
 
4-Bromo-2,6-dimethoxybenzaldehyde
Catalog # APOSOR350031-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,6-dimethoxybenzaldehyde
Catalog # APOSOR350031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.07 g/mol
Storage Temperature:Ambient
CAS Number:1354050-38-6
Formula:C₉H₉BrO₃
MDL Number:MFCD22192498