2,5-Dimethylimidazo[1,2-a]pyridine 95+%
Catalog # APOSOR346512-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.189 g/mol
Storage Temperature:Ambient
CAS Number:6188-30-3
Formula:C₉H₁₀N₂
MDL Number:MFCD09263848