3-Bromomethyl-2,5-dichloropyridine 95+%
Catalog # APOSOR346477-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.913 g/mol
Storage Temperature:Ambient
CAS Number:1141990-05-7
Formula:C₆H₄BrCl₂N
MDL Number:MFCD16607532