(1,4-Diphenyl-1H-pyrrol-3-yl)-methanol
Catalog # APOSOR346308-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.307 g/mol
Storage Temperature:Ambient
CAS Number:1355334-89-2
Formula:C₁₇H₁₅NO
MDL Number:MFCD21362495