4,6-Dimethyl-pyrimidine-5-carboxylic acid methyl ester
Catalog # APOSOR346240-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.177 g/mol
Storage Temperature:Refrigerator
CAS Number:832090-44-5
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD11046670